Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-2-iodoaniline, 98%

Click to view available options
Quantity:
5g
25g
63069-48-7
C6H5ClIN
253.467
MFCD01863737
FLEJOBRWKBPUOX-UHFFFAOYSA-N
2-iodo-4-chloroaniline,4-chloro-2-iodo-aniline,4-chloro-2-iodo-phenylamine,4-chloro-2-iodophenylamine,benzenamine, 4-chloro-2-iodo,zlchem 532,pubchem3645,4-chloro-6-iodoaniline,acmc-1bcyk,2-iodo-4-chloro-aniline
2757615
4-chloro-2-iodoaniline
C1=CC(=C(C=C1Cl)I)N
63069-48-7
C6H5ClIN
253.467
MFCD01863737
FLEJOBRWKBPUOX-UHFFFAOYSA-N
2-iodo-4-chloroaniline,4-chloro-2-iodo-aniline,4-chloro-2-iodo-phenylamine,4-chloro-2-iodophenylamine,benzenamine, 4-chloro-2-iodo,zlchem 532,pubchem3645,4-chloro-6-iodoaniline,acmc-1bcyk,2-iodo-4-chloro-aniline
2757615
4-chloro-2-iodoaniline
C1=CC(=C(C=C1Cl)I)N
Quantity 5g
Flash Point >110°C (230°F)
Melting Point 41°C to 44°C
UN Number UN2811
Formula Weight 253.47
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-iodoaniline

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.