Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chloro-2-iodobenzoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13421-13-1
C7H4ClIO2
282.47
LRRDANNSUCQNDU-UHFFFAOYSA-N
4-chloro-2-iodobenzoicacid,2-iodo-4-chlorobenzoic acid,4-chloro-2-iodo-benzoic acid,benzoic acid, 4-chloro-2-iodo,buttpark 100\07-49,pubchem4018,acmc-209bu6,ksc519a3p,benzoic acid,4-chloro-2-iodo,4-chloro-2-iodobenzoic acid
139452
4-chloro-2-iodobenzoic acid
C1=CC(=C(C=C1Cl)I)C(=O)O
13421-13-1
C7H4ClIO2
282.47
LRRDANNSUCQNDU-UHFFFAOYSA-N
4-chloro-2-iodobenzoicacid,2-iodo-4-chlorobenzoic acid,4-chloro-2-iodo-benzoic acid,benzoic acid, 4-chloro-2-iodo,buttpark 100\07-49,pubchem4018,acmc-209bu6,ksc519a3p,benzoic acid,4-chloro-2-iodo,4-chloro-2-iodobenzoic acid
139452
4-chloro-2-iodobenzoic acid
C1=CC(=C(C=C1Cl)I)C(=O)O
Melting Point 165.2°C to 166.4°C
Infrared Spectrum Authentic
Water 3% max.
Color White
Quantity 5g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 282.47
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-iodobenzoic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning