Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-2-methyl-6-nitroaniline, 98%

Click to view available options
Quantity:
1g
5g
62790-50-5
C7H7ClN2O2
186.595
MFCD01320687
QDSCDFKGUAONPC-UHFFFAOYSA-N
4-chloro-2-methyl-6-nitrophenylamine,acmc-1b9ic,2-amino-5-chloro-3-nitrotoluene,2-methyl-4-chloro-6-nitroaniline,4-chloro-2-methyl-6-nitrobenzenamine,2-amino-5-chloro-3-methylnitrobenzene,benzenamine,4-chloro-2-methyl-6-nitro,benzenamine, 4-chloro-2-methyl-6-nitro
182355
4-chloro-2-methyl-6-nitroaniline
CC1=CC(=CC(=C1N)[N+](=O)[O-])Cl
62790-50-5
C7H7ClN2O2
186.595
MFCD01320687
QDSCDFKGUAONPC-UHFFFAOYSA-N
4-chloro-2-methyl-6-nitrophenylamine,acmc-1b9ic,2-amino-5-chloro-3-nitrotoluene,2-methyl-4-chloro-6-nitroaniline,4-chloro-2-methyl-6-nitrobenzenamine,2-amino-5-chloro-3-methylnitrobenzene,benzenamine,4-chloro-2-methyl-6-nitro,benzenamine, 4-chloro-2-methyl-6-nitro
182355
4-chloro-2-methyl-6-nitroaniline
CC1=CC(=CC(=C1N)[N+](=O)[O-])Cl
Quantity 5g
Melting Point 121°C to 123°C
Formula Weight 186.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-methyl-6-nitroaniline

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger