Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Chloro-2'-methylacetanilide, 97%

Click to view available options
Quantity:
10g
5202-86-8
C9H10ClNO
183.635
MFCD00027399
WMHHXYOPEQARIN-UHFFFAOYSA-N
4-chloro-2-methylacetanilide,n-4-chloro-2-methylphenyl acetamide,acetamide, n-4-chloro-2-methylphenyl,4'-chloro-2'-methylacetanilide,maybridge1_000114,4-chloro-2-methylaceta-nilide,acetamide,n-4-chloro-2-methylphenyl,n-4-chloro-2-methyl-phenyl-acetamide
78877
N-(4-chloro-2-methylphenyl)acetamide
CC1=C(C=CC(=C1)Cl)NC(=O)C
5202-86-8
C9H10ClNO
183.635
MFCD00027399
WMHHXYOPEQARIN-UHFFFAOYSA-N
4-chloro-2-methylacetanilide,n-4-chloro-2-methylphenyl acetamide,acetamide, n-4-chloro-2-methylphenyl,4'-chloro-2'-methylacetanilide,maybridge1_000114,4-chloro-2-methylaceta-nilide,acetamide,n-4-chloro-2-methylphenyl,n-4-chloro-2-methyl-phenyl-acetamide
78877
N-(4-chloro-2-methylphenyl)acetamide
CC1=C(C=CC(=C1)Cl)NC(=O)C
Quantity 10g
Melting Point 139°C to 140°C
Beilstein 2087943
Formula Weight 183.64
Percent Purity 97%
Chemical Name or Material 4'-Chloro-2'-methylacetanilide

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger