Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-2-methylanisole, 98%

Click to view available options
Quantity:
5g
25g
3260-85-3
C8H9ClO
156.609
MFCD00070771
VDYDAUQHTVCCBX-UHFFFAOYSA-N
4-chloro-2-methylanisole,5-chloro-2-methoxytoluene,unii-7g3kq2a26k,2-methyl-4-chloroanisole,labotest-bb ltbb003018,labotest-bb lt03497723,4-chloro-1-methoxy-2-methyl-benzene,4-chloro-2-methyl anisole,pubchem2665,benzene, 4-chloro-1-methoxy-2-methyl
76748
4-chloro-1-methoxy-2-methylbenzene
CC1=C(C=CC(=C1)Cl)OC
3260-85-3
C8H9ClO
156.609
MFCD00070771
VDYDAUQHTVCCBX-UHFFFAOYSA-N
4-chloro-2-methylanisole,5-chloro-2-methoxytoluene,unii-7g3kq2a26k,2-methyl-4-chloroanisole,labotest-bb ltbb003018,labotest-bb lt03497723,4-chloro-1-methoxy-2-methyl-benzene,4-chloro-2-methyl anisole,pubchem2665,benzene, 4-chloro-1-methoxy-2-methyl
76748
4-chloro-1-methoxy-2-methylbenzene
CC1=C(C=CC(=C1)Cl)OC
Flash Point 101°C (215°F)
Melting Point 32°C to 35°C
Quantity 25g
Formula Weight 156.61
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-methylanisole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning