Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-2-methylbenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
209919-30-2
C7H8BClO2
170.399
MFCD02683107
SRXXSLUUAWHGBZ-UHFFFAOYSA-N
4-chloro-2-methylbenzeneboronic acid,4-chloro-2-methylphenyl boronic acid,4-chloro-o-tolylboronic acid,4-chloro-o-tolueneboronic acid,4-chloro-2-methyl-phenyl boronic acid,2-methyl-4-chlorophenylboronic acid,2-methyl-4-chloro-phenylboronic acid,2-methyl-4-chlorophenyl-boronic acid,boronic acid, 4-chloro-2-methylphenyl
5017600
(4-chloro-2-methylphenyl)boronic acid
B(C1=C(C=C(C=C1)Cl)C)(O)O
209919-30-2
C7H8BClO2
170.399
MFCD02683107
SRXXSLUUAWHGBZ-UHFFFAOYSA-N
4-chloro-2-methylbenzeneboronic acid,4-chloro-2-methylphenyl boronic acid,4-chloro-o-tolylboronic acid,4-chloro-o-tolueneboronic acid,4-chloro-2-methyl-phenyl boronic acid,2-methyl-4-chlorophenylboronic acid,2-methyl-4-chloro-phenylboronic acid,2-methyl-4-chlorophenyl-boronic acid,boronic acid, 4-chloro-2-methylphenyl
5017600
(4-chloro-2-methylphenyl)boronic acid
B(C1=C(C=C(C=C1)Cl)C)(O)O
Melting Point 193°C to 195°C
Quantity 1g
Formula Weight 170.4
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-methylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning