Alfa Aesar™ 4-Chloro-2-(methylthio)pyrimidine, 98%

Manufacturer: Alfa Aesar™ B2148109

Quantity 10g

 View more versions of this product

Catalog No. AAB2148109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 49844-90-8
    Molecular Formula C5H5ClN2S
    Formula Weight 160.63
    MDL Number MFCD00006083
    UN Number UN1760
    Beilstein 118042
    Synonym 4-Chloro-2-(methylmercapto)pyrimidine
    Chemical Name or Material 4-Chloro-2-(methylthio)pyrimidine

    Specifications

    Formula Weight 160.63
    MDL Number MFCD00006083
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Recommended Storage −30° to −10°C
    Flash Point >110°C (230°F)
    Quantity 10g
    Molecular Formula C5H5ClN2S
    CAS 49844-90-8
    UN Number UN1760
    Beilstein 118042
    Synonym 4-Chloro-2-(methylmercapto)pyrimidine
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
    Boiling Point 118° to 120°C (9mmHg)
    Melting Point ≈−2°C
    Chemical Name or Material 4-Chloro-2-(methylthio)pyrimidine
    Assay 98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    −30° to −10°C

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit