Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-2-nitroaniline, 98%

Click to view available options
Quantity:
250g
1000g
5000g
89-63-4
C6H5ClN2O2
172.568
MFCD00007836
PBGKNXWGYQPUJK-UHFFFAOYSA-N
benzenamine, 4-chloro-2-nitro,p-chloro-o-nitroaniline,pcon,2-nitro-4-chloroaniline,red salt nbgl,devol red f,devol red salt f,red base ciba vi,red base irga vi,red salt ciba vi
6979
4-chloro-2-nitroaniline
C1=CC(=C(C=C1Cl)[N+](=O)[O-])N
89-63-4
C6H5ClN2O2
172.568
MFCD00007836
PBGKNXWGYQPUJK-UHFFFAOYSA-N
benzenamine, 4-chloro-2-nitro,p-chloro-o-nitroaniline,pcon,2-nitro-4-chloroaniline,red salt nbgl,devol red f,devol red salt f,red base ciba vi,red base irga vi,red salt ciba vi
6979
4-chloro-2-nitroaniline
C1=CC(=C(C=C1Cl)[N+](=O)[O-])N
Applications
4-Chloro-2-nitroaniline is used as pharmaceutical intermediate.

Solubility
Soluble in water (0.5 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Density 1.37
Odor Weak, Characteristic
Flash Point 191°C (375°F)
Quantity 1000g
Melting Point 114°C to 118°C
UN Number UN2237
Beilstein 512436
Solubility Information Soluble in water (0.5g/L).
Formula Weight 172.57
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-nitroaniline
Show More Show Less

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c

H300+H310+H330-H373-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROANILINES

EINECSNumber : 201-925-4

RTECSNumber : BX1575000

TSCA : Yes

Recommended Storage : Ambient temperatures