Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-2-nitroanisole, 99+%

Click to view available options
Quantity:
5g
25g
100g
89-21-4
C7H6ClNO3
187.579
MFCD00024327
OSAYFGJUEOYRHY-UHFFFAOYSA-N
4-chloro-2-nitroanisole,benzene, 4-chloro-1-methoxy-2-nitro,2-nitro-4-chloroanisole,anisole, 4-chloro-2-nitro,labotest-bb ltbb001192,labotest-bb lt03381711,4-methoxy-3-nitrochlorobenzene,4-chloro-2-nitro-1-methoxybenzene,4-chlor-2-nitroanisol,2-nitryl-4-chloroanisole
66631
4-chloro-1-methoxy-2-nitrobenzene
COC1=C(C=C(C=C1)Cl)[N+](=O)[O-]
89-21-4
C7H6ClNO3
187.579
MFCD00024327
OSAYFGJUEOYRHY-UHFFFAOYSA-N
4-chloro-2-nitroanisole,benzene, 4-chloro-1-methoxy-2-nitro,2-nitro-4-chloroanisole,anisole, 4-chloro-2-nitro,labotest-bb ltbb001192,labotest-bb lt03381711,4-methoxy-3-nitrochlorobenzene,4-chloro-2-nitro-1-methoxybenzene,4-chlor-2-nitroanisol,2-nitryl-4-chloroanisole
66631
4-chloro-1-methoxy-2-nitrobenzene
COC1=C(C=C(C=C1)Cl)[N+](=O)[O-]
Quantity 5g
Melting Point 97°C to 99°C
Formula Weight 187.58
Percent Purity ≥99%
Chemical Name or Material 4-Chloro-2-nitroanisole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning