Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-nitrobenzyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
938-71-6
C7H5Cl2NO2
206.03
OMPHLGROCARZOU-UHFFFAOYSA-N
4-chloro-2-nitrobenzyl chloride,4-chloro-1-chloromethyl-2-nitrobenzene,4-chloro-2-nitrobenzylchloride,acmc-20amlj,benzene, 4-chloro-1-chloromethyl-2-nitro,2-nitro-4-chlorobenzyl chloride,omphlgrocarzou-uhfffaoysa,.alpha.,4-dichloro-2-nitrotoluene
598399
4-chloro-1-(chloromethyl)-2-nitrobenzene
C1=CC(=C(C=C1Cl)[N+](=O)[O-])CCl
938-71-6
C7H5Cl2NO2
206.03
OMPHLGROCARZOU-UHFFFAOYSA-N
4-chloro-2-nitrobenzyl chloride,4-chloro-1-chloromethyl-2-nitrobenzene,4-chloro-2-nitrobenzylchloride,acmc-20amlj,benzene, 4-chloro-1-chloromethyl-2-nitro,2-nitro-4-chlorobenzyl chloride,omphlgrocarzou-uhfffaoysa,.alpha.,4-dichloro-2-nitrotoluene
598399
4-chloro-1-(chloromethyl)-2-nitrobenzene
C1=CC(=C(C=C1Cl)[N+](=O)[O-])CCl
Melting Point 38°C to 44°C
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 97.5% min. (GC)
Linear Formula ClC6H3(NO2)CH2Cl
Formula Weight 206.03
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-nitrobenzyl chloride

GHS Signal Word: Danger

EINECSNumber : 213-345-9