Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-2-nitrotoluene, 99%

Click to view available options
Quantity:
100g
500g
89-59-8
C7H6ClNO2
171.58
MFCD00007215
SQFLFRQWPBEDHM-UHFFFAOYSA-N
4-chloro-2-nitrotoluene,2-nitro-4-chlorotoluene,benzene, 4-chloro-1-methyl-2-nitro,4,2-chloronitrotoluene,p-chloro-o-nitrotoluene,toluene, 4-chloro-2-nitro,4-chloro-1-methyl-2-nitro-benzene,ccris 3116,4-chlor-2-nitrotoluol,pubchem18919
6975
4-chloro-1-methyl-2-nitrobenzene
CC1=C(C=C(C=C1)Cl)[N+](=O)[O-]
89-59-8
C7H6ClNO2
171.58
MFCD00007215
SQFLFRQWPBEDHM-UHFFFAOYSA-N
4-chloro-2-nitrotoluene,2-nitro-4-chlorotoluene,benzene, 4-chloro-1-methyl-2-nitro,4,2-chloronitrotoluene,p-chloro-o-nitrotoluene,toluene, 4-chloro-2-nitro,4-chloro-1-methyl-2-nitro-benzene,ccris 3116,4-chlor-2-nitrotoluol,pubchem18919
6975
4-chloro-1-methyl-2-nitrobenzene
CC1=C(C=C(C=C1)Cl)[N+](=O)[O-]
Boiling Point 242°C
Melting Point 34°C to 38°C
Flash Point 120°C (248°F)
Quantity 500g
UN Number UN3457
Formula Weight 171.58
Percent Purity 99%
Chemical Name or Material 4-Chloro-2-nitrotoluene

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROTOLUENES, SOLID

TSCA : TSCA