Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-(trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™

Catalog Number AAH34011MD
Click to view available options
Quantity:
250 mg
1 g
1261779-42-3
C8H3Cl2F3O2
259.005
MFCD18393813
ZYZNCCXCNJYIHJ-UHFFFAOYSA-N
4-chloro-2-trifluoromethoxy benzoyl chloride
66523565
4-chloro-2-(trifluoromethoxy)benzoyl chloride
C1=CC(=C(C=C1Cl)OC(F)(F)F)C(=O)Cl
1261779-42-3
C8H3Cl2F3O2
259.005
MFCD18393813
ZYZNCCXCNJYIHJ-UHFFFAOYSA-N
4-chloro-2-trifluoromethoxy benzoyl chloride
66523565
4-chloro-2-(trifluoromethoxy)benzoyl chloride
C1=CC(=C(C=C1Cl)OC(F)(F)F)C(=O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 259.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-(trifluoromethoxy)benzoyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit