Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-Chloro-2-(trifluoromethyl)aniline, 97%
Click to view available options
Quantity:
25g
100g
500g
Specifications
Refractive Index | 1.507 |
Density | 1.386 |
Melting Point | 8°C to 9°C |
Flash Point | 95°C (203°F) |
Boiling Point | 66°C to 67°C (3mmHg) |
Quantity | 500g |
UN Number | UN2810 |
Beilstein | 2366801 |
Formula Weight | 195.57 |
Percent Purity | 97% |
Show More |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H318-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 207-151-3
TSCA : Yes
Recommended Storage : Ambient temperatures