Alfa Aesar™ 4-Chloro-2-(trifluoromethyl)benzonitrile, 97%

Manufacturer: Alfa Aesar™ H2667506

Quantity 5g

 View more versions of this product

Catalog No. AAH2667506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 320-41-2
    Molecular Formula C8H3ClF3N
    Formula Weight 205.57
    MDL Number MFCD00060671
    UN Number UN3276
    Synonym 5-Chloro-2-cyanobenzotrifluoride
    Chemical Name or Material 4-Chloro-2-(trifluoromethyl)benzonitrile

    Specifications

    Formula Weight 205.57
    Odor Characteristic
    Molecular Formula C8H3ClF3N
    MDL Number MFCD00060671
    CAS 320-41-2
    UN Number UN3276
    Synonym 5-Chloro-2-cyanobenzotrifluoride
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    Quantity 5g
    Chemical Name or Material 4-Chloro-2-(trifluoromethyl)benzonitrile
    Assay 97%

    GHS H Statement
    H302-H312-H332-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit