Alfa Aesar™ 4-Chloro-2-(trifluoromethyl)benzyl bromide, 97%

Manufacturer: Alfa Aesar™ H2662006

Quantity 5g

 View more versions of this product

Catalog No. AAH2662006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 886496-75-9
    Molecular Formula C8H5BrClF3
    Formula Weight 273.48
    MDL Number MFCD04115840
    UN Number UN3265
    Chemical Name or Material 4-Chloro-2-(trifluoromethyl)benzyl bromide

    Specifications

    Formula Weight 273.48
    MDL Number MFCD04115840
    CAS 886496-75-9
    UN Number UN3265
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Quantity 5g
    Molecular Formula C8H5BrClF3
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 4-Chloro-2-(trifluoromethyl)benzyl bromide
    Assay 97%

    Description

    Applications
    4-Chloro-2-(trifluoromethyl)benzyl bromide is used in the synthesis of novel series of dithiocarbamates, via reaction with sodium salts of N,N-disubstituted dithiocarbamic acids

    Solubility
    Difficult to mix.

    Notes
    Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit