Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-2-(trifluoromethyl)pyridine, 98%

Click to view available options
Quantity:
250mg
1g
5g
131748-14-6
C6H3ClF3N
181.542
MFCD07368047
FZINIBTZNSPWQR-UHFFFAOYSA-N
4-chloro-2-trifluoromethyl pyridine,4-chloro-2-trifluoromethyl pyridine,2-trifluoromethyl-4-chloropyridine,pyridine,4-chloro-2-trifluoromethyl,pubchem15520,acmc-209bnu,4-chloro-2-trifluoromethyl pyridine,4-chloro-2-trifluoromethyl-pyridine,2-trifluoromethyl-4-chloropyridine,2-trifluoro methyl-4-chloro pyridine
14761442
4-chloro-2-(trifluoromethyl)pyridine
C1=CN=C(C=C1Cl)C(F)(F)F
131748-14-6
C6H3ClF3N
181.542
MFCD07368047
FZINIBTZNSPWQR-UHFFFAOYSA-N
4-chloro-2-trifluoromethyl pyridine,4-chloro-2-trifluoromethyl pyridine,2-trifluoromethyl-4-chloropyridine,pyridine,4-chloro-2-trifluoromethyl,pubchem15520,acmc-209bnu,4-chloro-2-trifluoromethyl pyridine,4-chloro-2-trifluoromethyl-pyridine,2-trifluoromethyl-4-chloropyridine,2-trifluoro methyl-4-chloro pyridine
14761442
4-chloro-2-(trifluoromethyl)pyridine
C1=CN=C(C=C1Cl)C(F)(F)F
Applications
Chloro(trifluoromethyl)pyridines derivatives are used as model substrates for regioexhaustive functionalization.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Color Colorless
Quantity 250mg
UN Number UN2810
Formula Weight 181.54
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)pyridine

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.