Alfa Aesar™ 4-Chloro-3,5-difluoropyridine, 97%

Manufacturer: Alfa Aesar™ H31611-03

Quantity 1g

 View more versions of this product

Catalog No. AAH3161103

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 851178-97-7
    Molecular Formula C5H2ClF2N
    Formula Weight 149.53
    MDL Number MFCD09998885
    UN Number UN1993
    Chemical Name or Material 4-Chloro-3,5-difluoropyridine

    Specifications

    Formula Weight 149.53
    Molecular Formula C5H2ClF2N
    MDL Number MFCD09998885
    UN Number UN1993
    Flash Point 28.8°C(84°F)
    Quantity 1g
    CAS 851178-97-7
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H302-H312-H332-H315-H319-H335
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    Boiling Point 124°C (760mmHg)
    Chemical Name or Material 4-Chloro-3,5-difluoropyridine
    Assay 97%

    GHS H Statement
    H226-H302-H312-H332-H315-H319-H335
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit