Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-3,5-dimethylphenol, 98+%

Manufacturer:  Alfa Aesar™ B2196436

Quantity 500g

 View more versions of this product

Catalog No. AAB2196436


  Check Availability
Add to cart

Specifications

Description

Applications
It is used in hospitals and households for disinfection and sanitation. It is used in wound-cleansing applications and household antiseptics such as Dettol liquid, cream and ointments.

Solubility
Soluble in benzene, toluene, glycerin, chloroform, acetone, isopropanol, alcohol, ether, terpenes and in solutions of alkali hydroxides. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, strong bases.
Description & Specifications

Chemical Identifiers

88-04-0
156.61
1862539
4-Chloro-3,5-xylenol; PCMX
C8H9ClO
MFCD00002324
14,2176
4-Chloro-3,5-dimethylphenol

Specifications

138°C (280°F)
112° to 116°C
88-04-0
C8H9ClO
MFCD00002324
14,2176
4-Chloro-3,5-dimethylphenol
≈246°C
500g
Odorless
156.61
1862539
4-Chloro-3,5-xylenol; PCMX
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P362-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : ZE6850000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit