Alfa Aesar™ 4-Chloro-3,5-dinitrobenzotrifluoride, 97%

Manufacturer: Alfa Aesar™ A1044622

Quantity 100g

 View more versions of this product

Catalog No. AAA1044622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 393-75-9
    Molecular Formula C7H2ClF3N2O4
    Formula Weight 270.55
    MDL Number MFCD00007068
    UN Number UN2811
    Beilstein 1220937
    Chemical Name or Material 4-Chloro-3,5-dinitrobenzotrifluoride

    Specifications

    Formula Weight 270.55
    Health Hazard 3 GHS P Statement
    P261-P305+P351+P338-P361-P302+P350-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    IF ON SKIN: Gently wash with plenty of soap and water.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H2ClF3N2O4
    CAS 393-75-9
    UN Number UN2811
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H310-H302-H315-H319-H335
    Fatal in contact with skin.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number XS9065000
    Melting Point 53° to 58°C
    Quantity 100g
    TSCA TSCA
    MDL Number MFCD00007068
    Beilstein 1220937
    Flash Point 126°C (258°F)
    Boiling Point >250°C
    Chemical Name or Material 4-Chloro-3,5-dinitrobenzotrifluoride
    Assay 97%

    Description

    Applications
    4-Chloro-3,5-dinitrobenzotrifluoride is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

    Solubility
    Insoluble in water.

    Notes
    Incompatible with oxidizing agents, strong bases. Stable at room temperature.

    GHS H Statement
    H310-H302-H315-H319-H335
    Fatal in contact with skin.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P305+P351+P338-P361-P302+P350-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    IF ON SKIN: Gently wash with plenty of soap and water.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit