Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chloro-3-cyanophenylboronic acid, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
871332-95-5
C7H5BClNO2
181.39
MFCD08056360
PRPLADNLEYNYFZ-UHFFFAOYSA-N
4-chloro-3-cyanophenyl boronic acid,5-borono-2-chlorobenzonitrile,4-chloro-3-cyanobenzeneboronic acid,4-chloro-3-cyano-phenyl boronic acid,4-chloro-3-cyanophenylboronicacid,pubchem17104,acmc-209qho,ksc447q0n,4-chloro-3-cyanobenzeneboronic acid 97+%
44119831
(4-chloro-3-cyanophenyl)boronic acid
B(C1=CC(=C(C=C1)Cl)C#N)(O)O
871332-95-5
C7H5BClNO2
181.39
MFCD08056360
PRPLADNLEYNYFZ-UHFFFAOYSA-N
4-chloro-3-cyanophenyl boronic acid,5-borono-2-chlorobenzonitrile,4-chloro-3-cyanobenzeneboronic acid,4-chloro-3-cyano-phenyl boronic acid,4-chloro-3-cyanophenylboronicacid,pubchem17104,acmc-209qho,ksc447q0n,4-chloro-3-cyanobenzeneboronic acid 97+%
44119831
(4-chloro-3-cyanophenyl)boronic acid
B(C1=CC(=C(C=C1)Cl)C#N)(O)O
Quantity 5g
Packaging Glass bottle
Color White to Yellow
Infrared Spectrum Authentic
Melting Point 300.0°C to 304.0°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 181.39
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material 4-chloro-3-cyanophenylboronic acid
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning