Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-fluoropyridine, 95%, Acros Organics

Click to view available options
Quantity:
5g
25g
2546-56-7
C5H3ClFN
131.54
BEQUUSCRAKEKQM-UHFFFAOYSA-N
pyridine, 4-chloro-3-fluoro,3-fluoro-4-chloro pyridine,3-fluoro-4-chloropyridine hydrochloride,pubchem6667,4-chloro-3fluoropyridine,3-fluoro-4-chloropyridine,acmc-209x6p,4-chloro-3-fluoro-pyridin,3-fluoro-4-chloro-pyridine,4-chloro-3-fluoro-pyridine
3852429
4-chloro-3-fluoropyridine
C1=CN=CC(=C1Cl)F
2546-56-7
C5H3ClFN
131.54
BEQUUSCRAKEKQM-UHFFFAOYSA-N
pyridine, 4-chloro-3-fluoro,3-fluoro-4-chloro pyridine,3-fluoro-4-chloropyridine hydrochloride,pubchem6667,4-chloro-3fluoropyridine,3-fluoro-4-chloropyridine,acmc-209x6p,4-chloro-3-fluoro-pyridin,3-fluoro-4-chloro-pyridine,4-chloro-3-fluoro-pyridine
3852429
4-chloro-3-fluoropyridine
C1=CN=CC(=C1Cl)F
Boiling Point 142°C to 143°C
Melting Point -27°C to 24°C
Flash Point 28°C
Quantity 5g
Assay Percent Range 94% min. (GC)
Formula Weight 131.54
Percent Purity 95%
Chemical Name or Material 4-Chloro-3-fluoropyridine

GHS Signal Word: Warning