Alfa Aesar™ 4-Chloro-3-fluoropyridine hydrochloride, 97%

Manufacturer: Alfa Aesar™ H2718414

Quantity 25g

 View more versions of this product

Catalog No. AAH2718414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 119229-74-2
    Molecular Formula C5H3ClFN·HCl
    Formula Weight 168
    MDL Number MFCD03453233
    Chemical Name or Material 4-Chloro-3-fluoropyridine hydrochloride

    Specifications

    Formula Weight 168
    Molecular Formula C5H3ClFN·HCl
    MDL Number MFCD03453233
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point ≈160°C (subl.)
    Quantity 25g
    CAS 119229-74-2
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 4-Chloro-3-fluoropyridine hydrochloride
    Assay 97%

    Description

    Applications
    4-Chloro-3-fluoropyridine hydrochloride is employed for the deprotection of phenol ethers.

    Solubility
    Soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit