Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-3-iodobenzotrifluoride, 98%

Click to view available options
Quantity:
25g
100g
672-57-1
C7H3ClF3I
306.451
MFCD00001037
SSLWFPKNYZEOTH-UHFFFAOYSA-N
1-chloro-2-iodo-4-trifluoromethyl benzene,4-chloro-3-iodobenzotrifluoride,3-iodo-4-chlorobenzotrifluoride,benzene, 1-chloro-2-iodo-4-trifluoromethyl,4-chloro-3-iodobenzo trifluoride,pubchem2714,attercop-chm at113459,1-chloro-2-iodo-4-trifluoromethyl-benzene,1-chloro-2-iodo-4-trifluoromethyl benzene #
69596
1-chloro-2-iodo-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)I)Cl
672-57-1
C7H3ClF3I
306.451
MFCD00001037
SSLWFPKNYZEOTH-UHFFFAOYSA-N
1-chloro-2-iodo-4-trifluoromethyl benzene,4-chloro-3-iodobenzotrifluoride,3-iodo-4-chlorobenzotrifluoride,benzene, 1-chloro-2-iodo-4-trifluoromethyl,4-chloro-3-iodobenzo trifluoride,pubchem2714,attercop-chm at113459,1-chloro-2-iodo-4-trifluoromethyl-benzene,1-chloro-2-iodo-4-trifluoromethyl benzene #
69596
1-chloro-2-iodo-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)I)Cl
Flash Point >110°C (230°F)
Boiling Point 58°C to 60°C (0.5mmHg)
Quantity 25g
Beilstein 1870212
Formula Weight 306.45
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-iodobenzotrifluoride

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning