Alfa Aesar™ 4-Chloro-3-methoxyaniline, 98%

Manufacturer: Alfa Aesar™ H2833103

Quantity 1g

 View more versions of this product

Catalog No. AAH2833103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13726-14-2
    Molecular Formula C7H8ClNO
    Formula Weight 157.6
    MDL Number MFCD00672967
    UN Number UN2233
    Synonym 4-Chloro-m-anisidine
    Chemical Name or Material 4-Chloro-3-methoxyaniline

    Specifications

    Formula Weight 157.6
    Odor Characteristic
    MDL Number MFCD00672967
    UN Number UN2233
    Molecular Formula C7H8ClNO
    CAS 13726-14-2
    Synonym 4-Chloro-m-anisidine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H373-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROANISIDINES
    Melting Point 78° to 81°C
    Quantity 1g
    Chemical Name or Material 4-Chloro-3-methoxyaniline
    Assay 98%

    GHS H Statement
    H301-H311-H373-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P301+P310-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit