Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-3-methylaniline, 98%

Click to view available options
Quantity:
5g
25g
7149-75-9
C7H8ClN
141.598
MFCD00066332
HIHCTGNZNHSZPP-UHFFFAOYSA-N
5-amino-2-chlorotoluene,benzenamine, 4-chloro-3-methyl,m-toluidine, 4-chloro,3-methyl-4-chloroaniline,4-chloro-m-toluidine,unii-6q2w6l7bwz,6q2w6l7bwz,4-chloro-3-methylphenylamine,4-chloro-3-methyl-phenylamine,4-chloro-3-methylphenyl amine
23536
4-chloro-3-methylaniline
CC1=C(C=CC(=C1)N)Cl
7149-75-9
C7H8ClN
141.598
MFCD00066332
HIHCTGNZNHSZPP-UHFFFAOYSA-N
5-amino-2-chlorotoluene,benzenamine, 4-chloro-3-methyl,m-toluidine, 4-chloro,3-methyl-4-chloroaniline,4-chloro-m-toluidine,unii-6q2w6l7bwz,6q2w6l7bwz,4-chloro-3-methylphenylamine,4-chloro-3-methyl-phenylamine,4-chloro-3-methylphenyl amine
23536
4-chloro-3-methylaniline
CC1=C(C=CC(=C1)N)Cl
Quantity 5g
Melting Point 82°C to 85°C
UN Number UN2239
Beilstein 636522
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-methylaniline

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROTOLUIDINES, SOLID