Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-3-methylbenzoyl chloride, 96%

Click to view available options
Quantity:
250mg
1g
21900-24-3
C8H6Cl2O
189.035
MFCD11111062
NECBSSKNQDXVKN-UHFFFAOYSA-N
3-methyl-4-chloro benzoylchloride,4-chloro-3-methylbenzoylchloride,3-methyl-4-chlorobenzoyl chloride,4-chloro-3-methyl-benzoyl chloride,4-chloranyl-3-methyl-benzoyl chloride,benzoyl chloride, 4-chloro-3-methyl
13058375
4-chloro-3-methylbenzoyl chloride
CC1=C(C=CC(=C1)C(=O)Cl)Cl
21900-24-3
C8H6Cl2O
189.035
MFCD11111062
NECBSSKNQDXVKN-UHFFFAOYSA-N
3-methyl-4-chloro benzoylchloride,4-chloro-3-methylbenzoylchloride,3-methyl-4-chlorobenzoyl chloride,4-chloro-3-methyl-benzoyl chloride,4-chloranyl-3-methyl-benzoyl chloride,benzoyl chloride, 4-chloro-3-methyl
13058375
4-chloro-3-methylbenzoyl chloride
CC1=C(C=CC(=C1)C(=O)Cl)Cl
Color Colorless to Yellow
Quantity 250mg
Sensitivity Moisture Sensitive
UN Number UN3265
Formula Weight 189.03
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 4-Chloro-3-methylbenzoyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.