Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-methylbenzylamine hydrochloride, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
1264198-68-6
C8H9N·ClH
192.09
WVNIGUUQWGGELM-UHFFFAOYSA-N
4-chloro-3-methylbenzylamine hydrochloride,4-chloro-3-methylphenyl methanamine hydrochloride,4-chloro-3-methyl-benzenemethanamine hydrochloride
110210981
(4-chloro-3-methylphenyl)methanamine;hydrochloride
CC1=C(C=CC(=C1)CN)Cl.Cl
1264198-68-6
C8H9N·ClH
192.09
WVNIGUUQWGGELM-UHFFFAOYSA-N
4-chloro-3-methylbenzylamine hydrochloride,4-chloro-3-methylphenyl methanamine hydrochloride,4-chloro-3-methyl-benzenemethanamine hydrochloride
110210981
(4-chloro-3-methylphenyl)methanamine;hydrochloride
CC1=C(C=CC(=C1)CN)Cl.Cl
Color Undesignated
Packaging Glass Bottle
Infrared Spectrum Authentic
Quantity 1g
Melting Point 252.3°C to 253.5°C
Assay Percent Range 96% min. (HPLC)
Formula Weight 192.09
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-methylbenzylamine hydrochloride

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning