Alfa Aesar™ 4-Chloro-3-methylphenol, 98%

Manufacturer: Alfa Aesar™ 36637A1

Quantity 1kg

 View more versions of this product

Catalog No. AA36637A1

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 59-50-7
    Formula Weight 142.59
    MDL Number MFCD00002323
    Merck Index 14,2133
    Synonym 4-Chloro-m-cresol
    Chemical Name or Material 4-Chloro-3-methylphenol

    Specifications

    Formula Weight 142.59
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H311-H318-H302-H317
    Toxic in contact with skin.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    MDL Number MFCD00002323
    Synonym 4-Chloro-m-cresol
    Flash Point 118° (244°F)
    Melting Point 65° to 68°C
    Quantity 1kg
    Merck Index 14,2133
    Health Hazard 1 Danger
    Boiling Point 235°C
    TSCA TSCA
    Odor Odorless
    Physical Form Powder
    CAS 59-50-7
    Chemical Name or Material 4-Chloro-3-methylphenol
    Assay 98%

    GHS H Statement
    H311-H318-H302-H317
    Toxic in contact with skin.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit