Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-3-methylphenol, 98%

Manufacturer:  Alfa Aesar™ 3663730

Quantity 250g

 View more versions of this product

Catalog No. AA3663730

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

59-50-7
MFCD00002323
4-Chloro-m-cresol
142.59
14,2133
4-Chloro-3-methylphenol

Specifications

118° (244°F)
65° to 68°C
59-50-7
Powder
MFCD00002323
4-Chloro-m-cresol
98%
235°C
250g
Odorless
142.59
14,2133
4-Chloro-3-methylphenol

Safety and Handling

GHS H Statement
H311-H318-H302-H317
Toxic in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit