Alfa Aesar™ 4-Chloro-3-methylphenyl isocyanate, 98%

Manufacturer: Alfa Aesar™ H3109909

Quantity 10g

 View more versions of this product

Catalog No. AAH3109909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 51488-20-1
    Molecular Formula C8H6ClNO
    Formula Weight 167.6
    MDL Number MFCD11101408
    UN Number UN2206
    Chemical Name or Material 4-Chloro-3-methylphenyl isocyanate

    Specifications

    Formula Weight 167.6
    MDL Number MFCD11101408
    CAS 51488-20-1
    UN Number UN2206
    Health Hazard 2 GHS H Statement
    H330-H334-H315-H319-H335
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
    Quantity 10g
    Molecular Formula C8H6ClNO
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P280h-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Chemical Name or Material 4-Chloro-3-methylphenyl isocyanate
    Assay 98%

    Description

    Applications
    It is employed as a intermediates for pharmaceutical.

    Solubility
    Hydrolyzes with water.

    Notes
    Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.

    GHS H Statement
    H330-H334-H315-H319-H335
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P260-P280h-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit