Alfa Aesar™ 4-Chloro-3-nitrobenzeneboronic acid pinacol ester, 98%

Manufacturer: Alfa Aesar™ H5302203

Quantity 1g

 View more versions of this product

Catalog No. AAH5302203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 913836-26-7
    Molecular Formula C12H15BClNO4
    Formula Weight 283.52
    MDL Number MFCD08235086
    Synonym 4-Chloro-3-nitrophenylboronic acid pinacol ester
    Chemical Name or Material 4-Chloro-3-nitrobenzeneboronic acid pinacol ester

    Specifications

    Formula Weight 283.52
    Molecular Formula C12H15BClNO4
    MDL Number MFCD08235086
    CAS 913836-26-7
    Synonym 4-Chloro-3-nitrophenylboronic acid pinacol ester
    Melting Point 56° to 58°C
    Quantity 1g
    Chemical Name or Material 4-Chloro-3-nitrobenzeneboronic acid pinacol ester
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit