Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitrobenzoic acid, 99%, Thermo Scientific™

Click to view available options
Quantity:
100g
500g
1000g
96-99-1
C7H4ClNO4
201.562
MFCD00007079
DFXQXFGFOLXAPO-UHFFFAOYSA-N
4-chloro-3-nitrobenzoic acid, 3-nitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-nitro, kyselina 4-chloro-3-nitrobenzoova, unii-0mlz915q9k, ccris 3126, kyselina 4-chloro-3-nitrobenzoova czech, 4-chloro-3-nitrobenzoic acic, 4-chloro-3-nitro-benzoic acid, 5-carboxy-2-chloronitrobenzene
7320
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Cl
96-99-1
C7H4ClNO4
201.562
MFCD00007079
DFXQXFGFOLXAPO-UHFFFAOYSA-N
4-chloro-3-nitrobenzoic acid, 3-nitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-nitro, kyselina 4-chloro-3-nitrobenzoova, unii-0mlz915q9k, ccris 3126, kyselina 4-chloro-3-nitrobenzoova czech, 4-chloro-3-nitrobenzoic acic, 4-chloro-3-nitro-benzoic acid, 5-carboxy-2-chloronitrobenzene
7320
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Density 1.645
Melting Point 180°C to 184°C
Odor Odorless
Beilstein 783626
Quantity 1000g
Formula Weight 201.57
Percent Purity 99%
Chemical Name or Material 4-Chloro-3-nitrobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 202-550-9

RTECSNumber : DG5425050

TSCA : Yes

Recommended Storage : Ambient temperatures

RUO – Research Use Only