Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitrobenzophenone 99%, Acros Organics

Click to view available options
Quantity:
100g
56107-02-9
C13H8ClNO3
261.661
MFCD00007081
YBDBYPQFIMSFJW-UHFFFAOYSA-N
4-chloro-3-nitrobenzophenone,methanone, 4-chloro-3-nitrophenyl phenyl,benzophenone, 4-chloro-3-nitro,4-chloro-3-nitrophenyl phenyl methanone,3-nitro-4-chlorobenzophenone,chembl53270,4-chloro-3-nitrophenyl-phenylmethanone,4-chloro-3-nitro-phenyl-phenyl-methanone,4-chloro-3-nitrophenyl phenyl ketone,acmc-20amlb
41743
(4-chloro-3-nitrophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)Cl)[N+](=O)[O-]
56107-02-9
C13H8ClNO3
261.661
MFCD00007081
YBDBYPQFIMSFJW-UHFFFAOYSA-N
4-chloro-3-nitrobenzophenone,methanone, 4-chloro-3-nitrophenyl phenyl,benzophenone, 4-chloro-3-nitro,4-chloro-3-nitrophenyl phenyl methanone,3-nitro-4-chlorobenzophenone,chembl53270,4-chloro-3-nitrophenyl-phenylmethanone,4-chloro-3-nitro-phenyl-phenyl-methanone,4-chloro-3-nitrophenyl phenyl ketone,acmc-20amlb
41743
(4-chloro-3-nitrophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)Cl)[N+](=O)[O-]
Color Yellow
Quantity 100g
Infrared Spectrum Authentic
Melting Point 101°C to 107°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ClC6H3(NO2)COC6H5
Beilstein 07, I, 230
Formula Weight 261.65
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 4-Chloro-3-nitrobenzophenone, 99%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.