Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-3-nitrobenzotrifluoride, 98%

Manufacturer:  Alfa Aesar™ A1148822

Quantity 100g

 View more versions of this product

Catalog No. AAA1148822


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

121-17-5
225.55
UN2307
4-Chloro-3-nitrobenzotrifluoride
C7H3ClF3NO2
MFCD00007084
523986

Specifications

101°C (213°F)
−3°C
121-17-5
225.55
UN2307
4-Chloro-3-nitrobenzotrifluoride
221° to 223°C
100g
C7H3ClF3NO2
MFCD00007084
523986
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 3-NITRO-4-CHLOROBENZOTRIFLUORIDE

RTECSNumber : XS9140000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit