Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-nitrobenzotrifluoride, 98%, Thermo Scientific™

Click to view available options
Quantity:
100g
500g
2500g
121-17-5
C7H3ClF3NO2
225.55
MFCD00007084
TZGFQIXRVUHDLE-UHFFFAOYSA-N
4-chloro-3-nitrobenzotrifluoride, 1-chloro-2-nitro-4-trifluoromethyl benzene, 3-nitro-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl nitrobenzene, benzene, 1-chloro-2-nitro-4-trifluoromethyl, cmnt, 2-nitro-4-trifluoromethylchlorobenzene, 3-nitro-4-chlorotrifluoromethylbenzene, 4-chloro-3-nitro-a,a,a-trifluorotoluene, 4-chloro-3-nitrobenzylidyne fluoride
8462
1-chloro-2-nitro-4-(trifluoromethyl)benzene
[O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F
121-17-5
C7H3ClF3NO2
225.55
MFCD00007084
TZGFQIXRVUHDLE-UHFFFAOYSA-N
4-chloro-3-nitrobenzotrifluoride, 1-chloro-2-nitro-4-trifluoromethyl benzene, 3-nitro-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl nitrobenzene, benzene, 1-chloro-2-nitro-4-trifluoromethyl, cmnt, 2-nitro-4-trifluoromethylchlorobenzene, 3-nitro-4-chlorotrifluoromethylbenzene, 4-chloro-3-nitro-a,a,a-trifluorotoluene, 4-chloro-3-nitrobenzylidyne fluoride
8462
1-chloro-2-nitro-4-(trifluoromethyl)benzene
[O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Density 1.535
Melting Point -3°C
Boiling Point 221°C to 223°C
Flash Point 101°C (213°F)
UN Number UN2307
Beilstein 523986
Refractive Index 1.49
Quantity 2500g
Formula Weight 225.55
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitrobenzotrifluoride
Show More Show Less

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 3-NITRO-4-CHLOROBENZOTRIFLUORIDE

EINECSNumber : 204-451-6

RTECSNumber : XS9140000

TSCA : Yes

Recommended Storage : Ambient temperatures

RUO – Research Use Only