Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chloro-3-nitrophenyl cyclopropyl ketone, ≥99%, Acros Organics

Click to view available options
Quantity:
2.5g
31545-26-3
C10H8ClNO3
225.628
MFCD00044820
UISHBVQQPVMSCS-UHFFFAOYSA-N
4-chloro-3-nitrophenyl cyclopropyl methanone,4-chloro-3-nitrophenyl cyclopropyl ketone,4-chloro-3-nitrophenyl-cyclopropylmethanone,4-chloranyl-3-nitro-phenyl-cyclopropyl-methanone
1268259
(4-chloro-3-nitrophenyl)-cyclopropylmethanone
C1CC1C(=O)C2=CC(=C(C=C2)Cl)[N+](=O)[O-]
31545-26-3
C10H8ClNO3
225.628
MFCD00044820
UISHBVQQPVMSCS-UHFFFAOYSA-N
4-chloro-3-nitrophenyl cyclopropyl methanone,4-chloro-3-nitrophenyl cyclopropyl ketone,4-chloro-3-nitrophenyl-cyclopropylmethanone,4-chloranyl-3-nitro-phenyl-cyclopropyl-methanone
1268259
(4-chloro-3-nitrophenyl)-cyclopropylmethanone
C1CC1C(=O)C2=CC(=C(C=C2)Cl)[N+](=O)[O-]
Melting Point 78°C to 80°C
Infrared Spectrum Authentic
Color Yellow
Quantity 2.5g
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Formula Weight 225.63
Physical Form Crystalline Powder
Percent Purity ≥99.0%
Chemical Name or Material 4-chloro-3-nitrophenyl cyclopropyl ketone, 99+%
Show More Show Less