Alfa Aesar™ 4-Chloro-3-nitrotoluene, 97+%

Manufacturer: Alfa Aesar™ A113830E

Quantity 2500g

 View more versions of this product

Catalog No. AAA113830E

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 89-60-1
    Molecular Formula C7H6ClNO2
    Formula Weight 171.58
    MDL Number MFCD00007085
    UN Number UN2433
    Beilstein 511055
    Synonym 1-Chloro-4-methyl-2-nitrobenzene
    Chemical Name or Material 4-Chloro-3-nitrotoluene

    Specifications

    Formula Weight 171.58
    Health Hazard 2 GHS H Statement
    H331-H373-H302-H312-H315-H319
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H6ClNO2
    CAS 89-60-1
    TSCA TSCA
    MDL Number MFCD00007085
    Synonym 1-Chloro-4-methyl-2-nitrobenzene
    Health Hazard 1 Danger
    Melting Point ≈7°C
    Quantity 2500g
    UN Number UN2433
    Flash Point 129°C (264°F)
    Boiling Point 260° to 262°C
    Beilstein 511055
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROTOLUENES, LIQUID
    Chemical Name or Material 4-Chloro-3-nitrotoluene
    Assay ≥97%

    GHS H Statement
    H331-H373-H302-H312-H315-H319
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit