Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-3-sulfamoylbenzoic acid, 98%

Click to view available options
Quantity:
50g
250g
1000g
1205-30-7
C7H6ClNO4S
235.638
MFCD00012375
FHQAWINGVCDTTG-UHFFFAOYSA-N
4-chloro-5-sulphamoylbenzoic acid,3-aminosulfonyl-4-chlorobenzoic acid,csba,sulfamido-3-chlorobenzoic acid,benzoic acid, 3-aminosulfonyl-4-chloro,3-sulfamoyl-4-chlorobenzoic acid,benzoic acid, 4-chloro-3-sulfamoyl,4-chloro-3-sulfamoyl-benzoic acid,4-chloro-5-sulfamoylbenzoic acid,4-chloro-3-sulphamoylbenzoic acid
14568
4-chloro-3-sulfamoylbenzoic acid
C1=CC(=C(C=C1C(=O)O)S(=O)(=O)N)Cl
1205-30-7
C7H6ClNO4S
235.638
MFCD00012375
FHQAWINGVCDTTG-UHFFFAOYSA-N
4-chloro-5-sulphamoylbenzoic acid,3-aminosulfonyl-4-chlorobenzoic acid,csba,sulfamido-3-chlorobenzoic acid,benzoic acid, 3-aminosulfonyl-4-chloro,3-sulfamoyl-4-chlorobenzoic acid,benzoic acid, 4-chloro-3-sulfamoyl,4-chloro-3-sulfamoyl-benzoic acid,4-chloro-5-sulfamoylbenzoic acid,4-chloro-3-sulphamoylbenzoic acid
14568
4-chloro-3-sulfamoylbenzoic acid
C1=CC(=C(C=C1C(=O)O)S(=O)(=O)N)Cl
Quantity 50g
Melting Point 255°C to 260°C
Formula Weight 235.65
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-sulfamoylbenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning