Alfa Aesar™ 4-Chloro-3-(trifluoromethoxy)aniline, 97%

Manufacturer: Alfa Aesar™ H3175203

Quantity 1g

 View more versions of this product

Catalog No. AAH3175203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 97608-50-9
    Molecular Formula C7H5ClF3NO
    Formula Weight 211.57
    MDL Number MFCD09832255
    UN Number UN2810
    Chemical Name or Material 4-Chloro-3-(trifluoromethoxy)aniline

    Specifications

    Formula Weight 211.57
    Molecular Formula C7H5ClF3NO
    MDL Number MFCD09832255
    UN Number UN2810
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Quantity 1g
    CAS 97608-50-9
    Chemical Name or Material 4-Chloro-3-(trifluoromethoxy)aniline
    Assay 97%

    Description

    Applications
    4-Chloro-3-(trifluoromethoxy)aniline is widely utilized as an intermediate in organic synthesis.

    Notes
    Store in a cool and dry place. Keep the container tightly sealed.

    GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit