Alfa Aesar™ 4-Chloro-3-(trifluoromethoxy)benzyl bromide, 97%

Manufacturer: Alfa Aesar™ H2625803

Quantity 1g

 View more versions of this product

Catalog No. AAH2625803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 886500-93-2
    Molecular Formula C8H5BrClF3O
    Formula Weight 289.48
    MDL Number MFCD06660249
    UN Number UN3265
    Chemical Name or Material 4-Chloro-3-(trifluoromethoxy)benzyl bromide

    Specifications

    Formula Weight 289.48
    Molecular Formula C8H5BrClF3O
    MDL Number MFCD06660249
    CAS 886500-93-2
    UN Number UN3265
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Quantity 1g
    Chemical Name or Material 4-Chloro-3-(trifluoromethoxy)benzyl bromide
    Assay 97%

    Description

    Applications
    4-Chloro-3-(trifluoromethoxy)benzyl bromide is used as a pharmaceutical intermediate.

    Solubility
    Not miscible or difficult to mix with water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit