Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-(trifluoromethyl)benzenesulfonyl chloride, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
32333-53-2
C7H3Cl2F3O2S
279.06
SSULGNXFUGLULI-UHFFFAOYSA-N
4-chloro-3-trifluoromethyl benzenesulfonyl chloride,4-chloro-3-trifluoromethyl benzene-1-sulfonyl chloride,4-chloro-3-trifluoromethyl-benzenesulfonyl chloride,4-chloro-3-trifluoromethyl benzenesulfonylchloride,4-chloro-3-trifluoromethyl benzenesulphonyl chloride,4-chloro-5-trifluoromethyl benzenesulfonyl chloride,benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl,ksc222e0j,4-chloro-3-trifluoromethylbenzenesulfonylchloride,4-chloro-3-trifluoromethyl benzenesulfonyl chlori
2780719
4-chloro-3-(trifluoromethyl)benzenesulfonyl chloride
C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)Cl
32333-53-2
C7H3Cl2F3O2S
279.06
SSULGNXFUGLULI-UHFFFAOYSA-N
4-chloro-3-trifluoromethyl benzenesulfonyl chloride,4-chloro-3-trifluoromethyl benzene-1-sulfonyl chloride,4-chloro-3-trifluoromethyl-benzenesulfonyl chloride,4-chloro-3-trifluoromethyl benzenesulfonylchloride,4-chloro-3-trifluoromethyl benzenesulphonyl chloride,4-chloro-5-trifluoromethyl benzenesulfonyl chloride,benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl,ksc222e0j,4-chloro-3-trifluoromethylbenzenesulfonylchloride,4-chloro-3-trifluoromethyl benzenesulfonyl chlori
2780719
4-chloro-3-(trifluoromethyl)benzenesulfonyl chloride
C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)Cl
Quantity 5g
Melting Point 28.0°C to 30.0°C
Assay Percent Range 96% min. (GC)
Formula Weight 279.06
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzenesulfonyl chloride

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi