Alfa Aesar™ 4-Chloro-4'-fluorobutyrophenone, 96%

Manufacturer: Alfa Aesar™ A1112122

Quantity 100g

 View more versions of this product

Catalog No. AAA1112122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3874-54-2
    Molecular Formula C10H10ClFO
    Formula Weight 200.64
    MDL Number MFCD00001007
    Beilstein 608741
    Synonym 3-Chloropropyl 4-fluorophenyl ketone
    Chemical Name or Material 4-Chloro-4'-fluorobutyrophenone

    Specifications

    Formula Weight 200.64
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 3874-54-2
    Beilstein 608741
    Synonym 3-Chloropropyl 4-fluorophenyl ketone
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Quantity 100g
    TSCA TSCA
    Recommended Storage 2°C to 8°C
    Flash Point 152°C (306°F)
    Boiling Point 120° to 122°C (0.7mmHg)
    Molecular Formula C10H10ClFO
    MDL Number MFCD00001007
    Health Hazard 1 Warning
    Melting Point 5° to 6°C
    Chemical Name or Material 4-Chloro-4'-fluorobutyrophenone
    Assay 96%

    GHS H Statement
    H302
    Harmful if swallowed.

    2°C to 8°C

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit