Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-4'-hydroxybenzophenone, 97%

Click to view available options
Quantity:
10g
42019-78-3
C13H9ClO2
232.663
MFCD00002357
RUETVLNXAGWCDS-UHFFFAOYSA-N
4-chloro-4'-hydroxybenzophenone,4-chlorophenyl 4-hydroxyphenyl methanone,4-hydroxy-4'-chlorobenzophenone,4-p-chlorobenzoylphenol,4-4-chlorobenzoyl phenol,4-chlorophenyl 4-hydroxyphenyl-methanone,methanone, 4-chlorophenyl 4-hydroxyphenyl,4-chlorophenyl-4-hydroxyphenyl methanone,4-chlorophenyl 4-hydroxyphenyl ketone,pubchem3317
123504
(4-chlorophenyl)-(4-hydroxyphenyl)methanone
C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)Cl)O
42019-78-3
C13H9ClO2
232.663
MFCD00002357
RUETVLNXAGWCDS-UHFFFAOYSA-N
4-chloro-4'-hydroxybenzophenone,4-chlorophenyl 4-hydroxyphenyl methanone,4-hydroxy-4'-chlorobenzophenone,4-p-chlorobenzoylphenol,4-4-chlorobenzoyl phenol,4-chlorophenyl 4-hydroxyphenyl-methanone,methanone, 4-chlorophenyl 4-hydroxyphenyl,4-chlorophenyl-4-hydroxyphenyl methanone,4-chlorophenyl 4-hydroxyphenyl ketone,pubchem3317
123504
(4-chlorophenyl)-(4-hydroxyphenyl)methanone
C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)Cl)O
Quantity 10g
Melting Point 177°C to 182°C
Boiling Point 256°C to 257°C (13mmHg)
Beilstein 2049956
Formula Weight 232.67
Percent Purity 97%
Chemical Name or Material 4-Chloro-4'-hydroxybenzophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning