Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Chloro-4'-methoxybutyrophenone, 97%

Click to view available options
Quantity:
5g
25g
40877-19-8
C11H13ClO2
212.673
MFCD00018995
NGBTWDPPZFGUAY-UHFFFAOYSA-N
4-chloro-4'-methoxybutyrophenone,4-chloro-1-4-methoxyphenyl butan-1-one,acmc-20aml7,gamma-chloro-4'-methoxybutyrophenone,4-chloro-1-4-methoxyphenyl-1-butanone,1-butanone,4-chloro-1-4-methoxyphenyl,4-chloro-1-4-methoxy-phenyl-butan-1-one,1-butanone, 4-chloro-1-4-methoxyphenyl,4-chloro-4 inverted exclamation marka-methoxybutyrophenone
96414
4-chloro-1-(4-methoxyphenyl)butan-1-one
COC1=CC=C(C=C1)C(=O)CCCCl
40877-19-8
C11H13ClO2
212.673
MFCD00018995
NGBTWDPPZFGUAY-UHFFFAOYSA-N
4-chloro-4'-methoxybutyrophenone,4-chloro-1-4-methoxyphenyl butan-1-one,acmc-20aml7,gamma-chloro-4'-methoxybutyrophenone,4-chloro-1-4-methoxyphenyl-1-butanone,1-butanone,4-chloro-1-4-methoxyphenyl,4-chloro-1-4-methoxy-phenyl-butan-1-one,1-butanone, 4-chloro-1-4-methoxyphenyl,4-chloro-4 inverted exclamation marka-methoxybutyrophenone
96414
4-chloro-1-(4-methoxyphenyl)butan-1-one
COC1=CC=C(C=C1)C(=O)CCCCl
Flash Point >110°C (230°F)
Quantity 5g
Boiling Point 159°C to 163°C (2mmHg)
Melting Point 30°C to 33°C
Beilstein 880054
Formula Weight 212.68
Percent Purity 97%
Chemical Name or Material 4-Chloro-4'-methoxybutyrophenone

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 255-125-5

TSCA : No

Recommended Storage : Ambient temperatures