Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-4'-methylbutyrophenone 95%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
38425-26-2
C11H13ClO
196.674
MFCD00018999
FUHGFUBJUUFMSF-UHFFFAOYSA-N
4-chloro-4'-methylbutyrophenone,4-chloro-1-4-methylphenyl butan-1-one,4-chloro-1-4-methylphenyl-1-oxobutane,1-butanone, 4-chloro-1-4-methylphenyl,4-chloro-1-p-tolyl butan-1-one,4'-methyl-4-chlorobutyrophenone,4-chloro-4'-methyl-butyrophenone,gamma-chloro-4'-methylbutyrophenone,4-chloro-4-methylbutyrophenone,4-chloro-1-4-methylphenyl-1-butanone
98745
4-chloro-1-(4-methylphenyl)butan-1-one
CC1=CC=C(C=C1)C(=O)CCCCl
38425-26-2
C11H13ClO
196.674
MFCD00018999
FUHGFUBJUUFMSF-UHFFFAOYSA-N
4-chloro-4'-methylbutyrophenone,4-chloro-1-4-methylphenyl butan-1-one,4-chloro-1-4-methylphenyl-1-oxobutane,1-butanone, 4-chloro-1-4-methylphenyl,4-chloro-1-p-tolyl butan-1-one,4'-methyl-4-chlorobutyrophenone,4-chloro-4'-methyl-butyrophenone,gamma-chloro-4'-methylbutyrophenone,4-chloro-4-methylbutyrophenone,4-chloro-1-4-methylphenyl-1-butanone
98745
4-chloro-1-(4-methylphenyl)butan-1-one
CC1=CC=C(C=C1)C(=O)CCCCl
Infrared Spectrum Authentic
Quantity 25g
Color Orange to Brown
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula CH3C6H4CO(CH2)3Cl
Formula Weight 196.68
Physical Form Crystalline Solid
Percent Purity ≥94%
Chemical Name or Material 4-Chloro-4';-methylbutyrophenone, 95%
Show More Show Less