Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-chloro-5-nitro-o-phenylenediamine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
100g
Packaging:
Glass bottle
67073-39-6
C6H6ClN3O2
187.59
MFCD03427594
LOQLMWFVXRZASN-UHFFFAOYSA-N
4-chloro-5-nitro-o-phenylenediamine,4-chloro-5-nitro-1,2-benzenediamine,5-chloro-4-nitrobenzene-1,2-diamine,4-chloro-5-nitro-1,2-phenylenediamine,4-chloro-5-nitro-benzene-1,2-diamine,1,2-benzenediamine, 4-chloro-5-nitro
5702618
4-chloro-5-nitrobenzene-1,2-diamine
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)N)N
67073-39-6
C6H6ClN3O2
187.59
MFCD03427594
LOQLMWFVXRZASN-UHFFFAOYSA-N
4-chloro-5-nitro-o-phenylenediamine,4-chloro-5-nitro-1,2-benzenediamine,5-chloro-4-nitrobenzene-1,2-diamine,4-chloro-5-nitro-1,2-phenylenediamine,4-chloro-5-nitro-benzene-1,2-diamine,1,2-benzenediamine, 4-chloro-5-nitro
5702618
4-chloro-5-nitrobenzene-1,2-diamine
C1=C(C(=CC(=C1[N+](=O)[O-])Cl)N)N
Packaging Glass bottle
Quantity 1g
Color Red
Melting Point 189.0°C to 193.0°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 187.59
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-chloro-5-nitro-o-phenylenediamine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning