Alfa Aesar™ 4-Chloro-5-trifluoromethyl-o-phenylenediamine, 95%

Manufacturer: Alfa Aesar™ H33761MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH33761MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 157590-59-5
    Formula Weight 210.59
    MDL Number MFCD00119550
    UN Number UN2811
    Synonym 2-Chloro-4,5-diaminobenzotrifluoride; 2-Amino-4-chloro-5-(trifluoromethyl)aniline
    Chemical Name or Material 4-Chloro-5-trifluoromethyl-o-phenylenediamine

    Specifications

    Formula Weight 210.59
    Melting Point 61° to 63°C
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00119550
    UN Number UN2811
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Physical Form Solid
    CAS 157590-59-5
    Synonym 2-Chloro-4,5-diaminobenzotrifluoride; 2-Amino-4-chloro-5-(trifluoromethyl)aniline
    Health Hazard 1 Danger
    Color White to brown
    Quantity 250mg
    Chemical Name or Material 4-Chloro-5-trifluoromethyl-o-phenylenediamine
    Assay 95%

    Description

    Applications
    4-Chloro-5-trifluoromethyl-o-phenylenediamine has application in dietary supplements, pharmaceutical or cosmeticeuticals.

    Solubility
    Slightly soluble in water.

    Notes
    Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit