Learn More
Accela Chembio IncSupplier Diversity Partner 4-CHLORO-6,7-DIHYDRO-5H-C 1G DFS Item
Manufacturer:  Accela Chembio IncSupplier Diversity Partner SY0116451G
4-chloro-6,7-dihydro-5h-cyclopentapyrimidine, 1g, 83942-13-6, MFCD12755922, 95%, Shelf Life: 1260 Days, Light Sensitive/nitrogen Or Argon/store In Freezer(-20) Upon Receipt
Catalog No. 50-481-101