Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-6-iodoquinazoline, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
98556-31-1
C8H4ClIN2
290.49
BDAIUOPDSRAOKI-UHFFFAOYSA-N
4-chloro-6-iodo-quinazoline,quinazoline, 4-chloro-6-iodo,6-iodo-4-chloroquinazoline,pubchem23075,4chloro-6-iodoquinazoline,6-iodo4-chloroquinazoline,4-chloro-6-iodo quinazoline,4-chloro-6-iodo-qulinazoline,6-iodine-4-chloroquine thiazole moiety
11173809
4-chloro-6-iodoquinazoline
C1=CC2=C(C=C1I)C(=NC=N2)Cl
98556-31-1
C8H4ClIN2
290.49
BDAIUOPDSRAOKI-UHFFFAOYSA-N
4-chloro-6-iodo-quinazoline,quinazoline, 4-chloro-6-iodo,6-iodo-4-chloroquinazoline,pubchem23075,4chloro-6-iodoquinazoline,6-iodo4-chloroquinazoline,4-chloro-6-iodo quinazoline,4-chloro-6-iodo-qulinazoline,6-iodine-4-chloroquine thiazole moiety
11173809
4-chloro-6-iodoquinazoline
C1=CC2=C(C=C1I)C(=NC=N2)Cl
Quantity 5g
Melting Point 175.0°C to 177.0°C
Assay Percent Range 96% min. (HPLC)
Formula Weight 290.49
Percent Purity 97%
Chemical Name or Material 4-Chloro-6-iodoquinazoline

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning