Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-6-methyl-2-methylthiopyrimidine, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
17119-73-2
C6H7ClN2S
174.65
ALMBOXQFPLQVLF-UHFFFAOYSA-N
4-chloro-6-methyl-2-methylthio pyrimidine,4-chloro-6-methyl-2-methylsulfanyl pyrimidine,4-chloro-6-methyl-2-methylsulfanyl-pyrimidine,pyrimidine, 4-chloro-6-methyl-2-methylthio,4-chloro-6-methyl-2-methylthiopyrimidine,2-methylmercapto-4-methyl-6-chloropyrimidine,pubchem21461,6-chloro-4-methyl-2-methylthiopyrimidine,4-chloro-6-methyl-2-methylthio-pyrimidine,2-methylthio-4-methyl-6-chloropyrimidine
241163
4-chloro-6-methyl-2-methylsulfanylpyrimidine
CC1=CC(=NC(=N1)SC)Cl
17119-73-2
C6H7ClN2S
174.65
ALMBOXQFPLQVLF-UHFFFAOYSA-N
4-chloro-6-methyl-2-methylthio pyrimidine,4-chloro-6-methyl-2-methylsulfanyl pyrimidine,4-chloro-6-methyl-2-methylsulfanyl-pyrimidine,pyrimidine, 4-chloro-6-methyl-2-methylthio,4-chloro-6-methyl-2-methylthiopyrimidine,2-methylmercapto-4-methyl-6-chloropyrimidine,pubchem21461,6-chloro-4-methyl-2-methylthiopyrimidine,4-chloro-6-methyl-2-methylthio-pyrimidine,2-methylthio-4-methyl-6-chloropyrimidine
241163
4-chloro-6-methyl-2-methylsulfanylpyrimidine
CC1=CC(=NC(=N1)SC)Cl
Quantity 5g
Infrared Spectrum Authentic
Melting Point 34.0°C to 37.0°C
Assay Percent Range 96% min. (GC)
Formula Weight 174.65
Percent Purity 97%
Chemical Name or Material 4-Chloro-6-methyl-2-methylthiopyrimidine

GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning